Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDPD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571536
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571536 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571536 Supplier Invitrogen™ Supplier No. PA571536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GDPD1 Polyclonal Antibody for Western Blot, IHC (P)

Hydrolyzes lysoglycerophospholipids to produce lysophosphatidic acid (LPA) and the corresponding amines. Shows a preference for 1-O-alkyl-sn-glycero-3-phosphocholine (lyso-PAF), lysophosphatidylethanolamine (lyso-PE) and lysophosphatidylcholine (lyso-PC). May be involved in bioactive N-acylethanolamine biosynthesis. Does not display glycerophosphodiester phosphodiesterase activity, since it cannot hydrolyze either glycerophosphoinositol or glycerophosphocholine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDPD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Gdpd1
Gene Accession No. Q8N9F7
Gene Alias 2610020H15Rik; GDE4; GDPD1; Glycerophosphodiester phosphodiesterase 4; glycerophosphodiester phosphodiesterase domain containing 1; glycerophosphodiester phosphodiesterase domain-containing protein 1; Lysophospholipase D GDPD1; RGD1311813; Unknown (protein for MGC:134023)
Gene Symbols Gdpd1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 18-46 amino acids from the N-terminal region of human GDPD1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284161
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.