Learn More
Invitrogen™ GDPD2 Monoclonal Antibody (501016)

Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | GDPD2 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 501016 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | GDPD2 |
| Gene Accession No. | Q9ESM6, Q9HCC8 |
| Gene Alias | 9130017L10Rik; EC 3.1.4.43; FLJ20207; Gde3; GDPD2; Glycerophosphodiester phosphodiesterase 3; glycerophosphodiester phosphodiesterase domain containing 2; glycerophosphodiester phosphodiesterase domain-containing protein 2; Glycerophosphoinositol inositolphosphodiesterase GDPD2; OBDPF; Osteoblast differentiation promoting factor; RGD1560780; UNQ1935/PRO4418 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.