Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDPD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572068
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572068 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572068 Supplier Invitrogen™ Supplier No. PA572068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GDPD2 Polyclonal Antibody for Western Blot

Has glycerophosphoinositol inositolphosphodiesterase activity and specifically hydrolyzes glycerophosphoinositol, with no activity for other substrates such as glycerophosphoinositol 4-phosphate, glycerophosphocholine, glycerophosphoethanolamine, and glycerophosphoserine. Accelerates the program of osteoblast differentiation and growth. May play a role in remodeling of the actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDPD2
Applications Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GDPD2
Gene Accession No. Q9HCC8
Gene Alias 9130017L10Rik; EC 3.1.4.43; FLJ20207; Gde3; GDPD2; Glycerophosphodiester phosphodiesterase 3; glycerophosphodiester phosphodiesterase domain containing 2; glycerophosphodiester phosphodiesterase domain-containing protein 2; Glycerophosphoinositol inositolphosphodiesterase GDPD2; OBDPF; Osteoblast differentiation promoting factor; RGD1560780; UNQ1935/PRO4418
Gene Symbols GDPD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 305-334 amino acids from the Central region of human GDPD2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54857
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.