Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDPD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113341
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113341 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113341 Supplier Invitrogen™ Supplier No. PA5113341
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GDPD2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Has glycerophosphoinositol inositolphosphodiesterase activity and specifically hydrolyzes glycerophosphoinositol, with no activity for other substrates such as glycerophosphoinositol 4-phosphate, glycerophosphocholine, glycerophosphoethanolamine, and glycerophosphoserine. Accelerates the program of osteoblast differentiation and growth. May play a role in remodeling of the actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDPD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene GDPD2
Gene Accession No. Q9HCC8
Gene Alias 9130017L10Rik; EC 3.1.4.43; FLJ20207; Gde3; GDPD2; Glycerophosphodiester phosphodiesterase 3; glycerophosphodiester phosphodiesterase domain containing 2; glycerophosphodiester phosphodiesterase domain-containing protein 2; Glycerophosphoinositol inositolphosphodiesterase GDPD2; OBDPF; Osteoblast differentiation promoting factor; RGD1560780; UNQ1935/PRO4418
Gene Symbols GDPD2
Host Species Rabbit
Immunogen Recombinant Human Glycerophosphoinositol inositolphosphodiesterase GDPD2 protein (210-490AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54857
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.