Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GDPD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564207
Description
Immunogen sequence: QLCLSCLTGI YGCRWKRYQR SHDDTTPWE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The glycerophosphodiester phosphodiesterase domain containing 5 (GDPD5) protein, also known as GDE2, is a seven transmembrane, widely expressed protein that is necessary for spinal motor neuron differentiation and retinoid-induced neuronal outgrowth. Altered choline phospholipid metabolism is a hallmark of cancer, and the elevated expression of GDPD5 correlates with malignant choline phospholipid metabolite profiles in human breast cancer.
Specifications
GDPD5 | |
Polyclonal | |
Unconjugated | |
GDPD5 | |
BC024955; GDE2; GDPD5; Glycerophosphodiester phosphodiesterase 2; glycerophosphodiester phosphodiesterase domain containing 5; glycerophosphodiester phosphodiesterase domain containing 5; glycerophosphodiester phosphodiesterase domain-containing protein 5; glycerophosphodiester phosphodiesterase domain-containing protein 5; PP1665; PP6037; PP9363; RGD1559673; UNQ1850/PRO3580 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
81544 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WTR4 | |
GDPD5 | |
Recombinant protein corresponding to Human GDPD5. Recombinant protein control fragment (Product #RP-103906). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction