Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDPD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564287
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564287 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564287 Supplier Invitrogen™ Supplier No. PA564287
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQKWRLGGIR SYNPEQIMLS AAVRRTSRDV SIMKEKLIFS EISDGVEVSD VLSVCSDNSY DTYANSTAT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

The glycerophosphodiester phosphodiesterase domain containing 5 (GDPD5) protein, also known as GDE2, is a seven transmembrane, widely expressed protein that is necessary for spinal motor neuron differentiation and retinoid-induced neuronal outgrowth. Altered choline phospholipid metabolism is a hallmark of cancer, and the elevated expression of GDPD5 correlates with malignant choline phospholipid metabolite profiles in human breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDPD5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GDPD5
Gene Accession No. Q8WTR4
Gene Alias BC024955; GDE2; GDPD5; Glycerophosphodiester phosphodiesterase 2; glycerophosphodiester phosphodiesterase domain containing 5; glycerophosphodiester phosphodiesterase domain containing 5; glycerophosphodiester phosphodiesterase domain-containing protein 5; glycerophosphodiester phosphodiesterase domain-containing protein 5; PP1665; PP6037; PP9363; RGD1559673; UNQ1850/PRO3580
Gene Symbols GDPD5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GDPD5. Recombinant protein control fragment (Product #RP-103685).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81544
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.