Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GDPD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111645
Description
Immunogen sequence: WWEVHNDYDE FNWYLYNRMG YWSD.
The glycerophosphodiester phosphodiesterase domain containing 5 (GDPD5) protein, also known as GDE2, is a seven transmembrane, widely expressed protein that is necessary for spinal motor neuron differentiation and retinoid-induced neuronal outgrowth. Altered choline phospholipid metabolism is a hallmark of cancer, and the elevated expression of GDPD5 correlates with malignant choline phospholipid metabolite profiles in human breast cancer.
Specifications
| GDPD5 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WTR4 | |
| GDPD5 | |
| Recombinant Protein corresonding to Human GDPD5. Recombinant protein control fragment (Product #RP-105467). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| Unconjugated | |
| GDPD5 | |
| BC024955; GDE2; GDPD5; Glycerophosphodiester phosphodiesterase 2; glycerophosphodiester phosphodiesterase domain containing 5; glycerophosphodiester phosphodiesterase domain containing 5; glycerophosphodiester phosphodiesterase domain-containing protein 5; glycerophosphodiester phosphodiesterase domain-containing protein 5; PP1665; PP6037; PP9363; RGD1559673; UNQ1850/PRO3580 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 81544 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction