Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDPD5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988904
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8904 Supplier Bioss Supplier No. BS9207R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The glycerophosphodiester phosphodiesterase domain containing 5 (GDPD5) protein, also known as GDE2, is a seven transmembrane, widely expressed protein that is necessary for spinal motor neuron differentiation and retinoid-induced neuronal outgrowth. Altered choline phospholipid metabolism is a hallmark of cancer, and the elevated expression of GDPD5 correlates with malignant choline phospholipid metabolite profiles in human breast cancer.

Specifications

Antigen GDPD5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GDPD5
Gene Accession No. Q8WTR4
Gene Alias BC024955; BOS_15000; Gde2; Gdpd5; Glycerophosphodiester phosphodiesterase 2; glycerophosphodiester phosphodiesterase domain containing 5; Glycerophosphodiester phosphodiesterase domain-containing protein 5; PP1665; PP6037; PP9363; RGD1559673; UNQ1850/PRO3580
Gene Symbols GDPD5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GDPD5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81544
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.