Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GDPGP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566282
Description
Immunogen sequence: VLLSLHPGFRV GFNSLGGLAS VNHLHLHGYY LAHRLPVEQA PSEPLDPGGH LHLLQDLPAP GFLFYTRGPG PDLESLISRV CRATDYLTDH E Highest antigen sequence identity to the following orthologs - mouse 87%, rat 23%.
Specific and highly efficient GDP-D-glucose phosphorylase regulating the levels of GDP-D-glucose in cells.
Specifications
GDPGP1 | |
Polyclonal | |
Unconjugated | |
GDPGP1 | |
C15orf58; D330012F22Rik; GDP-D-glucose phosphorylase 1; GDP-D-glucose phosphorylase C15orf58; GDP-D-glucose phosphorylase C15orf58 homolog; GDPGP1; RGD1560277; VTC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
390637 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZNW5 | |
GDPGP1 | |
Recombinant Human GDPGP1. Recombinant protein control fragment (Product #RP-106967). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction