Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDPGP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561684
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561684 Supplier Invitrogen™ Supplier No. PA561684
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IAHNLFVTRG APPGKTSPSS ALTGVRVILW ARKSSFGIKD GEAFNVALCE LAGHLPVKTS QDFSSLTEAA AVALIQDCRL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 26%.

Specific and highly efficient GDP-D-glucose phosphorylase regulating the levels of GDP-D-glucose in cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDPGP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GDPGP1
Gene Accession No. Q6ZNW5
Gene Alias C15orf58; D330012F22Rik; GDP-D-glucose phosphorylase 1; GDP-D-glucose phosphorylase C15orf58; GDP-D-glucose phosphorylase C15orf58 homolog; GDPGP1; RGD1560277; VTC2
Gene Symbols GDPGP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GDPGP1. Recombinant protein control fragment (Product #RP-98169).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390637
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.