Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Amersham™ AlkPhos Direct™ Labeling Modules

Catalog No. p-4907584
Click to view available options
Quantity:
2500 cm2 Membrane
This item is not returnable. View return policy
This item is not returnable. View return policy

Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes. AlkPhos Direct Labeling Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase.

DNA or RNA probes are labeled during a 30-minute reaction and resulting probes can be used without purification. Hybridization stringency can be controlled by temperature as well as salt concentration.

  • Hybridization stringency can be controlled by elevating temperatures up to 75°C
  • Detection of 60 fg target DNA (single-copy gene) in a genomic Southern blot
  • Rapid protocol saves two to three hours over conventional indirect methods

Order Info

Each system include the following reagents, sufficient for 25 or 50 labeling reactions and detection of 2500cm2 or 5000cm2 membrane (depending on product ordered): labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer and blocking agent

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description AlkPhos Direct Labeling and Detection Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase. CDP-Star or ECF detection can be used.
For Use With (Application) Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes
Includes Labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer and blocking agent
Product Type AlkPhos Direct Labeling Module
Quantity 2500 cm2 Membrane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.