Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester)
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Label or Dye:
Cy5
Cy7
Product Type:
Cy5 NHS Ester
Cy7 NHS Ester
Description
- Contains derivitized CyDyes that carry only one reactive group on each dye molecule
- pH range: 3 to 10
- DMSO tolerant
- Photostable
- Example protocols included with each dye pack

Specifications
Specifications
For Use With (Application) | For labeling via Amine groups, CyDyes are suitable for use in applications such as Fluorescence resonance energy transfer (FRET), time resolved-Fluorescence resonance energy transfer (TR-FRET) and Fluorescence polarization (FP) |
Quantity | 1 mg |
Product Type | Cy5 NHS Ester |
Kit Contents | Foil packs, each containing 1, 5, 10, 25 or 50mg dried dye |
pH Range | 3 to 10 |
Content And Storage | 2°C to 8°C |
Label or Dye | Cy5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction