Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Chelating Sepharose™ Fast Flow Medium

Catalog No. 45000113
Click to view available options
Quantity:
50 mL

Use for purifying native and recombinant proteins and peptides that have an affinity for metal ions. Chelating Sepharose™ Fast Flow Medium provides robust performance and regulatory support.

  • Immobilised Metal Ion Affinity chromatography media fulfills industrial demands for security of supply, robust performance and regulatory support
  • Supplied free of metal ions allowing possibility to charge with your metal ion of choice for optimized selectivity
  • Cross-linked 6% agarose beads modified with iminodiacetic immobilized to the base matrix by stable ether linkages and sufficiently long spacer arms

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 20% Ethanol, 8M Guanidine Hydrochloride, 8M Urea, 0.01M HCl, non-ionic Detergent, Acetate Buffer pH 4
Product Type Chelating Sepharose FF medium
Quantity 50 mL
Height (English) 9.84 in.
Height (Metric) 250mm
Ligand Type Iminodiacetic Acid
Max. Pressure 10psi
pH Range 3 to 13
Particle Size 90μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
For Use With (Application) Purifying native and recombinant Proteins and Peptides that have an affinity for Metal ions
Matrix Chelating Sepharose
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.