Learn More
Cytiva HiPrep™ HIC FF 16/10 Columns

Description
The 20mL HIC columns provide fast, preparative separation of proteins and other biomolecules. They are ready-to-use with four media choices: Phenyl Sepharose 6 Fast Flow (high sub); Phenyl Sepharose 6 Fast Flow (low sub); Butyl Sepharose 4 Fast Flow; and Octyl Sepharose 4 Fast Flow.
Features:
- Offer high capacity, excellent flow properties and reliable, reproducible separations
- Compatible with simple pump-based configurations or chromatography systems (e.g., AKTA)
Specifications (H2O at room temp.)
- Dimensions: 1.6 x 10cm
- Recommended flowrate: 2-10mL/min. (60-300cm/h)
- Maximum flowrate: 10mL/min. (300cm/hr.)
- Max. pressure over packed bed: 1.5bar (22psi, 0.15MPa)
- Column hardware pressure limit: 5bar (73psi, 0.5MPa)
- Storage: 20% ethanol
- Storage temperature: 4° to 30°C
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Buffer | 1M NaOH, 30% Isopropanol, 70% Ethanol, 6M Guanidine Hydrochloride, 30% Aceto nitrile, 1mM HCl |
Product Type | Octyl Sepharose |
Bed Volume | 20mL |
Diameter (English) Inner | 0.62 in. |
Diameter (Metric) Inner | 16mm |
Quantity | 1 x 20 mL |
Height (English) | 3.93 in. |
Height (Metric) | 100mm |
Flow Rate | >300 cm/hr. |
Material | Polypropylene |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.