Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Con A 4B Columns

Catalog No. p-4596136
Click to view available options
Quantity:
5 x 1mL
5 x 5mL
Diameter (English) Inner:
0.27 in.
0.62 in.
Flow Rate:
<20 mL/min.
<4 mL/min.

For separation and purification of glycoproteins, polysaccharides and glycolipids

  • Prepacked with Con A Sepharose™ 4B media
  • Bed volume: 1mL and 5mL
  • Bed dimensions: 7 x 25mm (1mL), 16 x 25mm (5mL)
  • Column material: Polypropylene (PP)
  • Max flow rate: 4mL/min (1mL), 20mL/min (5mL)
Media
  • Ligand: Concanavalin A (Con A)
  • Matrix: 4% agarose
  • Average particle size: 90μm
  • pH stability working range: 4 to 9

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffer
Product Type HiTrap Con A Sepharose 4B column
Bed Volume 1mL
Diameter (English) Inner 0.27 in.
Diameter (Metric) Inner 7mm
Quantity 5 x 1mL
Height (English) 0.98 in.
Height (Metric) 25mm
Flow Rate <4 mL/min.
Material Polypropylene
Ligand Type Concanavalin A (13mg Con A/mL drained medium)
Max. Pressure 3 bar
pH Range 4 to 9
Particle Size 45 to 165μm
Support Type 4% Agarose
Content And Storage 4° to 8°C
Column Type Ion Exchange Column
For Use With (Application) Convenient separation and purification of Glycoproteins, Polysaccharides and Glycolipids
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.