GE Healthcare Mono Q PC, Columns Prepacked with MonoBeads™ Available on GSA/VA Contract for Federal Government customers only.

Use Mono Q or S precision columns for fast, sensitive, and high-resolution ion exchange separations of biomolecules at micropreparative scale. GE Healthcare Life Sciences™ Mono Q or S PC, Columns Prepacked with MonoBeads™ achieve maximum resolution from the efficiency of small, perfectly spherical monodispersed particles.

Manufacturer: GE Healthcare 17517901

 View more versions of this product

Catalog No. 45-000-309

  Check Availability
Add to Cart

    Specifications

    Quantity Each
    Height (English) 3.93 in.
    Height (Metric) 100mm
    Ligand Type O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-N+ (CH3) 3
    Max. Pressure 40 bar
    Particle Size 10μm
    Diameter (Metric) Inner 4.6mm
    For Use With (Application) Small-scale polishing in purification of Proteins, Peptides and other biomolecules when high purity is required
    Flow Rate 0.05 to 3mL/min.
    Material PEEK
    pH Range 2 to 12
    Support Type Monodisperse porous Polystyrene/Divinyl Benzene bead
    Storage Requirements 4° to 30°C
    Bed Volume 1.7mL
    Diameter (English) Inner 0.18 in.
    Product Type Mono Q ion exchange column
    Buffer All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid

    • Maximum resolution achieved from efficiency of small spherical particles, optimally packed
    • Particles are coupled to Q or S ion exchangers with excellent sensitivities
    • Compatible with high-performance LC systems
    • Stable in all common buffers except high-concentration oxidizing and reactive agents, DMSO, DMF and formic acid

    • Tricorn columns are optimized for the polishing step when high purity is required
    • PC 1.6/5 columns are excellent for micropurification of subnanogram to microgram amounts of sample on high-performance LC systems
    • Mono Q: ionic capacity 0.27 to 0.37mmol Cl-/mL
    • Mono S: ionic capacity 0.12 to 0.15mmol Cl-/mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit