Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Nucleon™ BACC1 Genomic DNA Extraction Kit

Catalog No. 45000955
Click to view available options
Quantity:
25 Kits

Includes

Reagents A and B, 5.0M Sodium Perchlorate, and Nucleon resins.

Nucleon BACC DNA Extraction kits use a Nucleon resin to bind proteins ensuring high yields of high-quality, high molecular weight genomic DNA in only one hour

  • Phenol-free protocol requiring only 30 minutes to complete
  • Low-shearing protocol gives maximum recoveries of high-molecular weight DNA
  • For 50 preps of 1mL of whole blood or cultured cells (1 to 3 x 106 cells/mL)
  • Available in different formats/reaction scales (See 45-000-956 and 45-000-960)

TRUSTED_SUSTAINABILITY

Specifications

Description illustra Nucleon BACC system rapidly extracts high molecular weight DNA from whole blood and cultured cells.
Sample Type Blood, Cell Cultures
For Use With (Application) Rapid extraction of high-quality, high molecular weight genomic DNA from cell cultures and blood.
Includes Reagents A and B, 5M Sodium Perchlorate and Nucleon™ Resins
Product Type illustra Nucleon BACC1
Quantity 25 Kits
Yield < 12 μg DNA/mg tissue
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.