Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GEF-H1 Mouse anti-Human, Unconjugated, Clone: 3C5, MilliporeSigma™

Catalog No. MABC10962MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC10962MI 25 μg
MABC10961MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC10962MI Supplier MilliporeSigma Supplier No. MABC109625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GEF-H1 Monoclonal antibody specifically detects GEF-H1 in Human samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GEF-H1
Applications Western Blot
Classification Monoclonal
Clone 3C5
Conjugate Unconjugated
Gene Accession No. NP_001155855
Gene Alias Rho guanine nucleotide exchange factor 2;Guanine nucleotide exchange factor H1;Microtubule-regulated Rho-GEF;Proliferating cell nucleolar antigen p40
Gene Symbols ARHGEF2, KIAA0651, LFP40
Host Species Mouse
Immunogen A linear peptide from the N-terminal region of human GEF-H1
Molecular Weight of Antigen ∼111 kDa observed; 111.54 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 3C5 is a mouse monoclonal antibody that specifically detects human Rho guanine nucleotide exchange factor 2 (GEF-H1).
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.