Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GEF-H1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578490 Supplier Invitrogen™ Supplier No. PA578490
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: CG10188 over expression fruit fly Predicted reactivity: Drosophila (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GEF-H1 activates Rho GTPases by promoting the exchange of GDP and GTP. Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. GEF-H1 may be involved in epithelial barrier permeability, cell motility and polarization, dendritic spine morphology, antigen presentation, leukemic cell differentiation, cell cycle regulation, innate immune response, and cancer. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GEF-H1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene CG10188
Gene Accession No. 0
Gene Alias AA408978; ARHGEF2; CG10188; CG10188-PA; CG10188-PB; CG10188-PC; cyst; cyst-PA; cyst-PB; cyst-PC; cysts; DKFZp547L106; DKFZp547P1516; Dmel\CG10188; Dmel_CG10188; FLJ45440; GEF; GEFH1; GEF-H1; Guanine nucleotide exchange factor H1; Kiaa0651; Lbcl1; LBC'S first cousin; Lfc; LFP40; Lymphoid blast crisis-like 1; microtubule-regulated Rho-GEF; mKIAA0651; oncogene LFC; OTTHUMP00000018855; P40; Proliferating cell nucleolar antigen p40; rho guanine nucleotide exchange factor 2; rho guanine nucleotide exchange factor 2-like protein; Rho/Rac guanine nucleotide exchange factor (GEF) 2; Rho/Rac guanine nucleotide exchange factor 2; rhobin
Gene Symbols CG10188
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of Drosophila GEF-H1. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 35237
Target Species Drosophila
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.