Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GEF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560645 Supplier Invitrogen™ Supplier No. PA560645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGEERKRELQ VPEENRADSQ DEKSQTFLGK SEEVTGKQED HGIKEKGVPV SGQEAKEPES WDGGRLGAVG RARSREEENE HHGPSMPALI APEDSPHCDL FP Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 30%.

This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GEF5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGEF5
Gene Accession No. Q12774
Gene Alias 2210412D05Rik; AA717842; ARH5; ARHGEF5; AW495314; DKFZp686N1969; Ephexin 3; Ephexin-3; GEF5; Guanine nucleotide regulatory protein TIM; oncogene TIM; P60; p60 TIM; Rho guanine nucleotide exchange factor (GEF) 5; Rho guanine nucleotide exchange factor 5; TIM; TIM1; Transforming immortalized mammary oncogene
Gene Symbols ARHGEF5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GEF5. Recombinant protein control fragment (Product #RP-104368).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7984
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.