Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GEFT Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306354AF594
Description
GEFT Polyclonal specifically detects GEFT in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GEFT | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
GEFTRAC/CDC42 exchange factor, Guanine nucleotide exchange factor GEFT, p63RhoGEFrho guanine nucleotide exchange factor 25, Rac/Cdc42/Rho exchange factor GEFT, Rho guanine nucleotide exchange factor (GEF) 25, RhoA/RAC/CDC42 exchange factor, RhoA/Rac/Cdc42 guanine nucleotide exchange factor GEFT | |
Produced in rabbits immunized with E. coli-derived Human GEFT fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
115557 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction