Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GEFT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544047
Description
Peptide sequence: RLGDQAQGST HTPINGSLPS LLLSPKGEVA RALLPLDKQA LGDIPQAPHD Sequence homology: Cow: 92%; Dog: 85%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Mouse: 85%; Pig: 100%; Rabbit: 85%; Rat: 92%.
The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange. The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange (Souchet et al., 2002 ).
Specifications
GEFT | |
Polyclonal | |
Unconjugated | |
Arhgef25 | |
2410008H17Rik; ARHGEF25; D10Ertd610e; Geft; guanine nucleotide exchange factor GEFT; p63RhoGEF; RAC/CDC42 exchange factor; RAC/CDC42 exchange factor isoform 2; rac/Cdc42/Rho exchange factor GEFT; Rho guanine nucleotide exchange factor (GEF) 25; Rho guanine nucleotide exchange factor 25; RhoA/RAC/CDC42 exchange factor; rhoA/Rac/Cdc42 guanine nucleotide exchange factor GEFT | |
Rabbit | |
Affinity chromatography | |
RUO | |
115557 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86VW2 | |
Arhgef25 | |
Synthetic peptide directed towards the C-terminal of human GEFT. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction