Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GEFT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106412 Supplier Invitrogen™ Supplier No. PA5106412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GEFT.

The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange. The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange (Souchet et al., 2002 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GEFT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Arhgef25
Gene Accession No. Q6P720, Q86VW2, Q9CWR0
Gene Alias 2410008H17Rik; ARHGEF25; D10Ertd610e; Geft; guanine nucleotide exchange factor GEFT; p63RhoGEF; RAC/CDC42 exchange factor; RAC/CDC42 exchange factor isoform 2; rac/Cdc42/Rho exchange factor GEFT; Rho guanine nucleotide exchange factor (GEF) 25; Rho guanine nucleotide exchange factor 25; RhoA/RAC/CDC42 exchange factor; rhoA/Rac/Cdc42 guanine nucleotide exchange factor GEFT
Gene Symbols Arhgef25
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARHGEF25(Accession Q86VW2), corresponding to amino acid residues V323-S373.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115557, 314904, 52666
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.