Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GEFT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | GEFT |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
GEFT Polyclonal specifically detects GEFT in Human samples. It is validated for Western Blot.Specifications
| GEFT | |
| Polyclonal | |
| Rabbit | |
| Q86VW2 | |
| 115557 | |
| Synthetic peptides corresponding to GEFT(RhoA/RAC/CDC42 exchange factor) The peptide sequence was selected from the C terminal of GEFT. Peptide sequence RLGDQAQGSTHTPINGSLPSLLLSPKGEVARALLPLDKQALGDIPQAPHD. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| GEFTRAC/CDC42 exchange factor, Guanine nucleotide exchange factor GEFT, p63RhoGEFrho guanine nucleotide exchange factor 25, Rac/Cdc42/Rho exchange factor GEFT, Rho guanine nucleotide exchange factor (GEF) 25, RhoA/RAC/CDC42 exchange factor, RhoA/Rac/Cdc42 guanine nucleotide exchange factor GEFT | |
| ARHGEF25 | |
| IgG | |
| 64 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title