Learn More
Cytiva Gel Staining Tray Packs
Catalog No. 45002438 Shop All Cytiva Products
Change view
Click to view available options
Type:
Glass Plate Divider
For Use With (Equipment):
Hoefer SE 600 Vertical Unit
Catalog No. Type For Use With (Equipment)
45-002-438 Glass Plate Divider Hoefer SE 600 Vertical Unit
1 options
Catalog No. 45-002-438 Supplier Cytiva Supplier No. 80617918
Add to Cart
Add to Cart
For use with Hoefer Processor Plus fluid delivery system.
  • Club sandwich divider
  • Notched
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Pair two gel sandwiches so that up to four gels can be run at one time. 
For Use With (Equipment) Hoefer SE 600 Vertical Unit
Type Glass Plate Divider
Dimensions (L x W) 18 x 16 cm