Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Gelatin
SCI_ED

Catalog No. s25335 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Lab Grade

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 9000-70-8
MDL Number MFCD00081638
Synonym Absorbable Gelatin Sponge, Pharmagel.

Specifications

Density 0.68g/mL
Assay Percent Range 100% w/v
Packaging Lab Grade
Quantity 500 g
Grade Laboratory
Chemical Name or Material Gelatin
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.