Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelatin, 250 Bloom, Powder, NF, Spectrum™ Chemical
SDP

Catalog No. 18602894 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

GE105, 9000-70-8

Spectrum™ Chemical Gelatin, 250 Bloom, Powder, NF is used pharmaceutical formulations as thickening agent and emulsifier. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9000-70-8
MDL Number MFCD00081638

Specifications

CAS Max % 100%
Loss on Drying 15%
Packaging Fiber Drum
Quantity 45 kg
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.