Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelatin Type B (Laboratory), Fisher Chemical™

Catalog No. G73 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

CAS 9000-70-8
Molecular Formula Varies
MDL Number MFCD00081638
Synonym 100 Bloom, Gelfoam

Specifications

Color White
Packaging Poly Pail
Quantity 3 kg
Grade Laboratory
Physical Form Solid
Chemical Name or Material Gelatin Type B
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.