Learn More
Geldanamycin, 98+%
Description
Geldanamycin binds to and inhibits the cytosolic chaperone functions of heat shock protein 90 (HSP90). HSP90 maintains the stability and functional shape of many oncogenic signaling proteins. It interferes with conformational maturation of proteins and the cellular stress response. In addition, it is a potent inhibitor of the nuclear hormone receptor family.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Geldanamycin binds to and inhibits the cytosolic chaperone functions of heat shock protein 90 (HSP90). HSP90 maintains the stability and functional shape of many oncogenic signaling proteins. It interferes with conformational maturation of proteins and the cellular stress response. In addition, it is a potent inhibitor of the nuclear hormone receptor family.
Solubility
Soluble in dimethyl sulfoxide at 100 mg/mL.
Notes
Store in a cool dry place. Keep away from oxidizing agents. Protect from heat.

Specifications
Specifications
Type | Geldanamycin |
Physical Form | Powder |
Color | Orange to Yellow |
Quantity | 100 mg |
Solubility Information | Soluble in dimethyl sulfoxide at 100mg/mL. |
Formula Weight | 560.64 |
Percent Purity | ≥99% |
Chemical Name or Material | Geldanamycin |
Safety and Handling
RTECSNumber | LX8920000 |
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.