Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gelsolin/GSN Antibody (16H9), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326541100UL
Description
Gelsolin/GSN Monoclonal antibody specifically detects Gelsolin/GSN in Human samples. It is validated for Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
| Gelsolin/GSN | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunocytochemistry | |
| 16H9 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type) | |
| A synthesized peptide derived from Human Gelsolin/GSN [UniProt P06396] | |
| 100 μL | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Tumor Suppressors | |
| 2934 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction