Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelsolin/GSN Antibody (20) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$243.50 - $560.50

Specifications

Antigen Gelsolin/GSN
Clone 20
Concentration 1.0 mg/mL
Dilution Western Blot 1:100-1:2000, ELISA 1:4000, Immunohistochemistry 1:6000, Immunocytochemistry/Immunofluorescence 1:2000 Immunohistochemistry-Paraffin 1:6000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018333
SDP
View Documents
Novus Biologicals
NBP1051610.025MG
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NBP105161
SDP
View Documents
Novus Biologicals
NBP105161
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

Gelsolin/GSN Monoclonal specifically detects Gelsolin/GSN in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Gelsolin/GSN
1.0 mg/mL
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot, ELISA
Unconjugated
Mouse
Human
Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type)
GSN
IgG1 κ
Protein A or G purified
Gelsolin (human)
20
Western Blot 1:100-1:2000, ELISA 1:4000, Immunohistochemistry 1:6000, Immunocytochemistry/Immunofluorescence 1:2000 Immunohistochemistry-Paraffin 1:6000
Monoclonal
Purified
RUO
P06396
2934
Gelsolin purified from human plasma
Primary
Store at 4C in the dark.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.