Learn More
Invitrogen™ Gelsolin Monoclonal Antibody (GT1656)

Description
Positive Control: HeLa, rat plasma, mouse plasma Predicted Reactivity: Chicken (80%), Sheep (96%), Bovine (95%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Gelsolin |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | GT1656 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | GSN |
| Gene Accession No. | P06396, P13020, Q68FP1 |
| Gene Alias | Actin-depolymerizing factor; ADF; ADFBrevin; AGEL; Brevin; DKFZp313L0718; gelsolin; gelsolin (amyloidosis, Finnish type); Gsb; Gsn; hypothetical protein LOC535077; plasma gelsolin precursor; RP11-477J21.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.