Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gelsolin Recombinant Rabbit Monoclonal Antibody (HL1931)
GREENER_CHOICE

Catalog No. PIMA547226
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547226 Supplier Invitrogen™ Supplier No. MA547226
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The protein encoded by this gene binds to the plus ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gelsolin
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL1931
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GSN
Gene Accession No. P06396
Gene Alias Actin-depolymerizing factor; ADF; ADFBrevin; AGEL; Brevin; DKFZp313L0718; gelsolin; gelsolin (amyloidosis, Finnish type); Gsb; Gsn; hypothetical protein LOC535077; plasma gelsolin precursor; RP11-477J21.1
Gene Symbols GSN
Host Species Rabbit
Immunogen Recombinant fragment of human Gelsolin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2934
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.