Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gelsolin Mouse anti-Human, Clone: 3C4A2, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 662801IG150UL

Description
The protein encoded by this gene binds to the "plus" ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.Specifications
Gelsolin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06396 | |
GSN | |
Gelsolin Fusion Protein Ag17916 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgA |
Immunohistochemistry (Paraffin), Western Blot | |
3C4A2 | |
Unconjugated | |
GSN | |
Actin depolymerizing factor, ADF, AGEL, Brevin, Gelsolin, GSN | |
Mouse | |
Affinity Chromatography | |
RUO | |
2934 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction