Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GEM Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP158906
Description
GEM Polyclonal specifically detects GEM in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
GEM | |
Polyclonal | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
GTP binding protein overexpressed in skeletal muscle, GTP-binding mitogen-induced T-cell protein, GTP-binding protein GEM, GTP-binding protein overexpressed in skeletal muscle, kinase-inducible Ras-like protein, KIRMGC26294, RAS-like protein KIR | |
Rabbit | |
33 kDa | |
100 μL | |
Signal Transduction | |
2669 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 4 - 8 ug/mL, Immunohistochemistry-Frozen | |
P55040 | |
GEM | |
Synthetic peptides corresponding to GEM(GTP binding protein overexpressed in skeletal muscle) The peptide sequence was selected from the C terminal of GEM (NP_005252). Peptide sequence ETSAAVQHNVKELFEGIVRQVRLRRDSKEKNERRLAYQKRKESMPRKARRFW The peptide sequence for this immunogen was taken from within the described region. | |
Protein A purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Chicken: 92%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction