Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GEM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109761
Description
Immunogen sequence: MTLNNVTMRQ GTVGMQPQQQ RWSIPADGRH LMVQKEPHQY SHRNRHSATP EDHCRRSWSS DSTDSVISSE SGNTYYRVVL IGEQGVGKST LANIFAGVHD SMDSDCEVLG EDTYERTLMV DGESATIILL DMWENKGENE WLHDHCMQVG.
GEM belongs to the RAD/GEM family of GTP-binding proteins. It is associated with the inner face of the plasma membrane and could play a role as a regulatory protein in receptor-mediated signal transduction.The protein encoded by this gene belongs to the RAD/GEM family of GTP-binding proteins. It is associated with the inner face of the plasma membrane and could play a role as a regulatory protein in receptor-mediated signal transduction. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.
Specifications
GEM | |
Polyclonal | |
Unconjugated | |
GEM | |
AV020497; Gem; GTP binding protein (gene overexpressed in skeletal muscle); GTP binding protein overexpressed in skeletal muscle; GTP-binding mitogen-induced T-cell protein; GTP-binding protein GEM; GTP-binding protein overexpressed in skeletal muscle; kinase-inducible Ras-like protein; KIR; MGC26294; RAS-like protein KIR | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2669 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.73 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P55040 | |
GEM | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GEM (NP_005252.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction