Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Gemcitabine hydrochloride
GREENER_CHOICESDP

Catalog No. 325950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-595-0 50 mg
32-591-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-595-0 Supplier R&D Systems™ Supplier No. 3259/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. DNA synthesis inhibitor

Deoxycytidine analog that inhibits DNA synthesis. Metabolized to form gemcitabine triphosphate (dFdCTP) and gemcitabine diphosphate (dFdCDP). dFdCTD inhibits ribonucleotide reductase causing a reduction in cellular nucleotides. dFdCTP is incorporated in DNA resulting in DNA strand termination. Displays antitumor activity in vitro and in vivo.

Chemical Identifiers

CAS 122111-03-9
Molecular Formula C9H12ClF2N3O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 299.66
Purity >99%
Molecular Weight (g/mol) 299.659
Chemical Name or Material Gemcitabine hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.