Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gemfibrozil, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602896
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Poly Pail

G1076, 25812-30-0, C15H22O3

Spectrum™ Chemical Gemfibrozil, USP is used to reduce cholesterol and triglycerides (fatty acids) in the blood. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 25812-30-0
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number MFCD00079335
InChI Key HEMJJKBWTPKOJG-UHFFFAOYSA-N
IUPAC Name 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid
SMILES CC1=CC=C(C)C(OCCCC(C)(C)C(O)=O)=C1

Specifications

CAS Max % 100%
Quantity 25 g
Packaging Amber Glass Bottle
Percent Purity 98 to 102%
Grade USP
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.