Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gemin 1/SMA Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981477
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1477 Supplier Bioss Supplier No. BS11561R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The SMN1 gene is part of a 500 kb inverted duplication on chromosome 5q13. This duplicated region contains at least four genes and repetitive elements which make it prone to rearrangements and deletions. The repetitiveness and complexity of the sequence have also caused difficulty in determining the organization of this genomic region. The telomeric and centromeric copies of this gene are nearly identical and encode the same protein - survival motor neuron protein. The SMN complex plays a catalyst role in the assemble of small nuclear ribonucleoproteins, the building blocks of the spliceosome. Mutations in the SMN1 gene are known to cause spinal muscular atrophy 1/2.

Specifications

Antigen Gemin 1/SMA
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SMN1
Gene Accession No. Q16637
Gene Alias BCD541; Component of gems 1; GEMIN1; Gemin-1; SMA; SMA@; SMA1; SMA2; SMA3; SMA4; SMN; SMN1; SMN2; SMNC; SMNT; survival motor neuron 1 protein; Survival motor neuron protein; survival of motor neuron 1, telomeric; T-BCD541; TDRD16A; tudor domain containing 16A
Gene Symbols SMN1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Gemin 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6606
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.