Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gemin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566055 Supplier Invitrogen™ Supplier No. PA566055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARRCSEVRLLV DSKDDERVPA LNLLICLVSR YFDQRDLADE PS Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

SIP1 is one of the proteins found in the SMN complex, which consists of the survival of motor neuron (SMN) protein and several gemin proteins. The SMN complex is localized to a subnuclear compartment called gems (gemini of coiled bodies) and is required for assembly of spliceosomal snRNPs and for pre-mRNA splicing. SIP1 interacts directly with the SMN and it is required for formation of the SMN complex. A knockout mouse targeting the mouse homolog of this gene exhibited disrupted snRNP assembly and motor neuron degeneration. However, knockdown of the SIP1 mRNA in motor neurons showed normal motor axons while that of SMN mRNA did show abnormal motor axon outgrowth, indicating that SIP1 may have additional roles outside of the SMN complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gemin 2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GEMIN2
Gene Accession No. O14893
Gene Alias 1700012N19Rik; Component of gems 2; gem (nuclear organelle) associated protein 2; gem nuclear organelle associated protein 2; gem nuclear organelle associated protein 2 S homeolog; gem-associated protein 2; GEMIN2; Gemin-2; gemin2.S; SIP 1; SIP1; SIP-1; SIP1 delta; sip1-delta; SMN interacting protein 1-delta; SMN interacting protein-1; SMN-interacting protein 1; survival of motor neuron protein interacting protein 1; Survival of motor neuron protein-interacting protein 1; survivor of motor neuron protein interacting protein 1; XELAEV_18042638mg
Gene Symbols GEMIN2
Host Species Rabbit
Immunogen Recombinant Human Gemin 2. Recombinant protein control fragment (Product #RP-105060).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8487
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.