Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gemin 3 Antibody (SR1015), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Gemin 3 |
---|---|
Clone | SR1015 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
Gemin 3 Monoclonal antibody specifically detects Gemin 3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
Gemin 3 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
Component of gems 3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 20, DEAD box protein 20, DEAD box protein DP 103, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20, 103kD, DKFZp434H052, DP103DEAD-box protein DP103, EC 3.6.4.13, Gemin-3, GEMIN3gemin-3, probable ATP-dependent RNA helicase DDX20, SMN-interacting protein | |
A synthesized peptide derived from human Gemin 3 (Uniprot #: Q9UHI6) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1015 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
11218 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title