Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Gemin 3 Monoclonal Antibody (1C9F6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531714
Description
MA5-31714 has been tested in indirect ELISA.
Gemin 3 is a member of the DEAD box protein family which are characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD) and are putative RNA helicases. DEAD box proteins such as Gemin 3 are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of DEAD box family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Gemin 3 has an ATPase activity and a component of the survival of motor neurons (SMN) complex. The Gemin 3 protein may play a catalytic role in the function of the SMN complex on RNPs. Diseases associated with Gemin 3 protein dysfunction include spinal muscular dystrophy and muscular atrophy.
Specifications
Gemin 3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9UHI6 | |
Ddx20 | |
Purified recombinant fragment of human DDX20 (AA: 725-824) expressed in E. coli. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
1C9F6 | |
Unconjugated | |
Ddx20 | |
component of gems 3; DDX 20; DDX20; DEAD (Asp-Glu-Ala-Asp) box polypeptide 20; DEAD box protein 20; DEAD box protein DP 103; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20, 103kD; DEAD/H BOX 20; DEAD-box helicase 20; DEAD-box protein DP103; DKFZP434H052; DP 103; Dp103; GEMIN3; Gemin-3; I79_021005; probable ATP-dependent RNA helicase DDX20; putative ATP-dependent RNA helicase DDX20; regulator of steroidogenic factor 1; ROSF-1; SMN-interacting protein | |
Mouse | |
Protein G | |
RUO | |
11218, 84473 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction