Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gemin 3 Monoclonal Antibody (7B2B1)
GREENER_CHOICE

Catalog No. PIMA531713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531713 Supplier Invitrogen™ Supplier No. MA531713
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31713 has been tested in indirect ELISA.

Gemin 3 is a member of the DEAD box protein family which are characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD) and are putative RNA helicases. DEAD box proteins such as Gemin 3 are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of DEAD box family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Gemin 3 has an ATPase activity and a component of the survival of motor neurons (SMN) complex. The Gemin 3 protein may play a catalytic role in the function of the SMN complex on RNPs. Diseases associated with Gemin 3 protein dysfunction include spinal muscular dystrophy and muscular atrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gemin 3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7B2B1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Ddx20
Gene Accession No. Q9UHI6
Gene Alias component of gems 3; DDX 20; DDX20; DEAD (Asp-Glu-Ala-Asp) box polypeptide 20; DEAD box protein 20; DEAD box protein DP 103; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20, 103kD; DEAD/H BOX 20; DEAD-box helicase 20; DEAD-box protein DP103; DKFZP434H052; DP 103; Dp103; GEMIN3; Gemin-3; I79_021005; probable ATP-dependent RNA helicase DDX20; putative ATP-dependent RNA helicase DDX20; regulator of steroidogenic factor 1; ROSF-1; SMN-interacting protein
Gene Symbols Ddx20
Host Species Mouse
Immunogen Purified recombinant fragment of human DDX20 (AA: 725-824) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11218
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.