Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gemin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564346
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564346 Supplier Invitrogen™ Supplier No. PA564346
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQGTSYDSYR LCDSLTSFSQ NATLYLNRTS LSKEDRQVVS ELAECVRDFL RKTSTVLKNR ALEDITASIA MAVIQQKMDR HMEVCYIFAS EKKWAFSDEW V Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.

The product of this gene is part of a large complex localized to the cytoplasm, nucleoli, and to discrete nuclear bodies called Gemini bodies. The complex functions in spliceosomal snRNP assembly in the cytoplasm, and regenerates spliceosomes required for pre-mRNA splicing in the nucleus. The encoded protein directly interacts with a DEAD box protein and several spliceosome core proteins. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gemin 4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GEMIN4
Gene Accession No. P57678
Gene Alias 4932415L08Rik; AU015359; component of gems 4; DKFZp434B131; DKFZp434D174; gem (nuclear organelle) associated protein 4; gem nuclear organelle associated protein 4; gem-associated protein 4; GEMIN4; Gemin-4; HC56; HCAP1; HCC-associated protein 1; HHRF-1; p97; RGD1563715
Gene Symbols GEMIN4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Gemin 4. Recombinant protein control fragment (Product #RP-104285).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50628
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.