Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gemin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588031 Supplier Invitrogen™ Supplier No. PA588031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LREISSAAAH SQPFAWKKKA LIIIWAKVLQ PHPVTPSDTE TRWQEDLFFS VGNMIPTINH TILFELLKSL EASGLFIQLL MALPTTICHA ELERFLEHVT VDTSAEDVAF FLDVWWEVMK HKGHPQDPLL SQFSAMAHKY LPALDEFPHP PKRLRSDPDA CPTMPLLAML LRGLTQIQSR ILGPGRKCCA LANLADMLTV FALTEDDPQE VSATVYLDKL ATVISVWNSD TQNPYHQQAL AEKVKEAERD VSLTSLAKLP SETIFVGCEF LHHLLREWGE ELQAVLRSSQ GTSYDSYRLC D; Positive Samples: HeLa, MCF-7, SH-SY5Y, Jurkat, Mouse brain, Mouse kidney, Mouse liver; Cellular Location: Cytoplasm, Nucleus, gem, nucleolus.

The product of this gene is part of a large complex localized to the cytoplasm, nucleoli, and to discrete nuclear bodies called Gemini bodies. The complex functions in spliceosomal snRNP assembly in the cytoplasm, and regenerates spliceosomes required for pre-mRNA splicing in the nucleus. The encoded protein directly interacts with a DEAD box protein and several spliceosome core proteins. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gemin 4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GEMIN4
Gene Accession No. P57678
Gene Alias 4932415L08Rik; AU015359; component of gems 4; DKFZp434B131; DKFZp434D174; gem (nuclear organelle) associated protein 4; gem nuclear organelle associated protein 4; gem-associated protein 4; GEMIN4; Gemin-4; HC56; HCAP1; HCC-associated protein 1; HHRF-1; p97; RGD1563715
Gene Symbols GEMIN4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-350 of human GEMIN4 (NP_0565362).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 276919, 50628
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.