Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gemin 6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557376 Supplier Invitrogen™ Supplier No. PA557376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNEGDHRVRE KLMHLFTSGD CKAYSPEDLE ERKNSLKKWL EKNHIPITEQ GDAPRTLCVA GVLTIDPPYG PENCSSS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.

GEMIN6 is part of a large macromolecular complex, localized to both the cytoplasm and the nucleus, that plays a role in the cytoplasmic assembly of small nuclear ribonucleoproteins (snRNPs). Other members of this complex include SMN (MIM 600354), GEMIN2 (SIP1; MIM 602595), GEMIN3 (DDX20; MIM 606168), GEMIN4 (MIM 606969), and GEMIN5 (MIM 607005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gemin 6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GEMIN6
Gene Accession No. Q8WXD5
Gene Alias 2610019B15Rik; 2810470M17Rik; FLJ23459; gem (nuclear organelle) associated protein 6; gem nuclear organelle associated protein 6; gem-associated protein 6; gemin 6; Gemin6; gemin-6; SIP2
Gene Symbols GEMIN6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Gemin 6. Recombinant protein control fragment (Product #RP-96142).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79833
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.