Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gemin 7 Monoclonal Antibody (4H6)
GREENER_CHOICE

Catalog No. PIMA522597
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522597 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522597 Supplier Invitrogen™ Supplier No. MA522597
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: ECPIAQESLE SQEQRARAAL RERYLRSLLA MVGHQVSFTL HEGVRVAAHF GATDLDVANF YVSQLQTPIG VQAEALLRCS DIISYTFKP.

The protein encoded by this gene is a component of the core SMN complex, which is required for pre-mRNA splicing in the nucleus. The encoded protein is found in the nucleoplasm, in nuclear gems (Gemini of Cajal bodies), and in the cytoplasm. Three transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gemin 7
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4H6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Gemin7
Gene Accession No. Q9H840
Gene Alias 2400008I04Rik; AI120175; AI461688; gem (nuclear organelle) associated protein 7; gem nuclear organelle associated protein 7; gem-associated protein 7; gemin 7; Gemin7; gemin-7; KIAA2024; SIP3; ZNF650
Gene Symbols Gemin7
Host Species Mouse
Immunogen GEMIN7 (NP_078983.1, 43 a.a. ~ 131 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79760
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.