Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Gemin 7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565700
Description
Immunogen sequence: GPDGFSRGFAP DGRRAPLRPE VPEIQECPIA QESLESQEQR ARAALRERYL RSLLAM Highest antigen sequence identity to the following orthologs - mouse 75%, rat 78%.
The protein encoded by this gene is a component of the core SMN complex, which is required for pre-mRNA splicing in the nucleus. The encoded protein is found in the nucleoplasm, in nuclear gems (Gemini of Cajal bodies), and in the cytoplasm. Three transcript variants encoding the same protein have been found for this gene.
Specifications
Gemin 7 | |
Polyclonal | |
Unconjugated | |
Gemin7 | |
2400008I04Rik; AI120175; AI461688; gem (nuclear organelle) associated protein 7; gem nuclear organelle associated protein 7; gem-associated protein 7; gemin 7; Gemin7; gemin-7; KIAA2024; SIP3; ZNF650 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79760 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H840 | |
Gemin7 | |
Recombinant Human Gemin 7. Recombinant protein control fragment (Product #RP-106372). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction