Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GEMIN2 Antibody (OTI4C11), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271374G
Description
GEMIN2 Monoclonal specifically detects GEMIN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
GEMIN2 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GEMIN2 | |
Full length recombinant protein of human GEMIN2 (NP_003607) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI4C11 | |
Western Blot, Immunohistochemistry | |
Component of gems 2, Gemin-2, GEMIN2survival of motor neuron protein-interacting protein 1, SIP1-delta, SMN interacting protein 1-delta, SMN-interacting protein 1, survival of motor neuron protein interacting protein 1 | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
8487.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction