Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GEMIN2 Antibody (SR1523), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32235650UL
Description
GEMIN2 Monoclonal antibody specifically detects GEMIN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
GEMIN2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoprecipitation | |
SR1523 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Component of gems 2, Gemin-2, GEMIN2survival of motor neuron protein-interacting protein 1, SIP1-delta, SMN interacting protein 1-delta, SMN-interacting protein 1, survival of motor neuron protein interacting protein 1 | |
A synthesized peptide derived from human GEMIN2 (Uniprot #: O14893) | |
50 μg | |
Core ESC Like Genes, Stem Cell Markers | |
8487 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction